Skip to Content
Merck
All Photos(1)

Documents

W372307

Sigma-Aldrich

2-Oxobutyric acid

≥95%, FG

Synonym(s):

2-Ketobutyric acid, α-Ketobutyric acid, Propionylformic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COCOOH
CAS Number:
Molecular Weight:
102.09
FEMA Number:
3723
Beilstein:
1700514
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.066
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

Assay

≥95%

bp

84 °C/20 mmHg (lit.)

mp

30-34 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

caramel; creamy; brown; sweet

storage temp.

2-8°C

SMILES string

CCC(=O)C(O)=O

InChI

1S/C4H6O3/c1-2-3(5)4(6)7/h2H2,1H3,(H,6,7)

InChI key

TYEYBOSBBBHJIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Oxobutyric acid is mainly found in the hydrolysates of proteins, reportedly being formed by the degradation of threonine.

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Probable Flavoring Principle in Vegetable?Protein Hydrolysates.
Sulser H, et al.
Journal of Food Science, 32(6), 611-615 (1967)
Review of thermally produced imitation meat flavors.
Wilson RA
Journal of Agricultural and Food Chemistry, 23(6), 1032-1037 (1975)
K Yaegaki et al.
Journal of periodontology, 63(9), 783-789 (1992-09-01)
The amounts of volatile sulfur compounds (VSC) and methyl mercaptan/hydrogen sulfide ratio in mouth air from patients with periodontal involvement were 8 times greater than those of control subjects. Our studies demonstrated that, in patients with periodontal disease: 1) the
Shufen Huang et al.
Journal of molecular biology, 396(3), 708-718 (2009-12-08)
In recent years, increased interest has been directed towards hydrogen sulfide (H2S) as the third gasotransmitter and its role in various diseases. Cystathionine-gamma-lyase (CSE) is one of the enzymes responsible for the endogenous production of H2S in mammals. With the
Chao Gao et al.
Applied and environmental microbiology, 76(5), 1679-1682 (2010-01-19)
2-Oxobutyrate is an important intermediate in the chemical, drug, and food industries. Whole cells of Pseudomonas stutzeri SDM, containing NAD-independent lactate dehydrogenases, effectively converted 2-hydroxybutyrate into 2-oxobutyrate. Under optimal conditions, the biocatalytic process produced 2-oxobutyrate at a high concentration (44.4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service