Skip to Content
Merck
All Photos(2)

Documents

K7125

Sigma-Aldrich

(±)-3-Methyl-2-oxovaleric acid sodium salt

Synonym(s):

(±)-Sodium 3-methyl-2-oxovalerate, 3-Methyl-2-oxopentanoic acid sodium salt, DL-α-Keto-β-methylvaleric acid sodium salt, Ketoisoleucine sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)COCOONa
CAS Number:
Molecular Weight:
152.12
Beilstein:
7779663
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

lipid type

saturated FAs

storage temp.

2-8°C

SMILES string

[Na+].CCC(C)C(=O)C([O-])=O

InChI

1S/C6H10O3.Na/c1-3-4(2)5(7)6(8)9;/h4H,3H2,1-2H3,(H,8,9);/q;+1/p-1

InChI key

SMDJDLCNOXJGKC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

(±)-3-Methyl-2-oxovaleric acid sodium salt is a mixture of the D- and L-3-methyl-2-oxovaleric acids. 3-methyl-2-oxovaleric acid is used as a substrate to study the specificity, distribution and kinetics of α-keto acid dehydrogenases. α-keto-β-methylvaleric acid is used to inhibit the mitochondrial α-ketoglutarate dehydrogenase complex (KGDHC) to induce and study nerve cell death. α-keto-β-methylvaleric acid may be studied as a reactive oxygen scavenger.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Lahnsteiner
Journal of fish biology, 75(4), 816-833 (2010-08-27)
The present study investigated (1) the free amino acid (FAA) composition in semen of rainbow trout Oncorhynchus mykiss and carp Cyprinus carpio, (2) enzyme systems involved in amino acid metabolism and (3) the effect of amino acids on sperm viability
Maritrini Colón et al.
PloS one, 6(1), e16099-e16099 (2011-01-27)
Gene duplication is a key evolutionary mechanism providing material for the generation of genes with new or modified functions. The fate of duplicated gene copies has been amply discussed and several models have been put forward to account for duplicate
Cláudia Funchal et al.
Journal of the neurological sciences, 260(1-2), 87-94 (2007-05-15)
Accumulation of the branched-chain alpha-keto acids (BCKA), alpha-ketoisocaproic acid (KIC), alpha-keto-beta-methylvaleric acid (KMV) and alpha-ketoisovaleric acid (KIV) and their respective branched-chain alpha-amino acids (BCAA) occurs in tissues and biological fluids of patients affected by the neurometabolic disorder maple syrup urine
Carmen Martínez-Cuesta et al.
Biotechnology letters, 25(8), 599-602 (2003-07-29)
The bacteriocin, lacticin 3147, increased isoleucine transamination by Lactococcus lactis IFPL359 in a cheese model system. The formation of alpha-keto-beta-methyl-n-valeric acid and 2-hydroxy-3-methyl-valeric acid increased by three times in cheese slurries at 12 degrees C and cheese aroma intensity increased
Hsueh-Meei Huang et al.
Annals of the New York Academy of Sciences, 1042, 272-278 (2005-06-21)
The alpha-ketoglutarate dehydrogenase complex (KGDHC) is a mitochondrial enzyme in the TCA cycle. Inhibition of KGDHC activity by alpha-keto-beta-methyl-n-valeric acid (KMV) is associated with neuron death. However, the effect of KMV in microglia is unclear. Therefore, we investigated the effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service