Skip to Content
Merck
All Photos(3)

Documents

520632

Sigma-Aldrich

Platinum(II) chloride

≥99.9% trace metals basis

Synonym(s):

Platinum dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PtCl2
CAS Number:
Molecular Weight:
265.98
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: platinum

impurities

≤1000.0 ppm Trace Metal Analysis

mp

581 °C (dec.) (lit.)

density

6.05 g/mL at 25 °C

SMILES string

Cl[Pt]Cl

InChI

1S/2ClH.Pt/h2*1H;/q;;+2/p-2

InChI key

CLSUSRZJUQMOHH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Platinum(II) chloride is a dark brown crystalline solid that exhibits high thermal stability and is widely used in catalysis and nanomaterial synthesis.

Application

Platinum(II) chloride can be used:
  • As a precursor to synthesize Pt nanoparticles.
  • To fabricate Ag-nanostructured grid transparent conductive electrodes.
  • As a catalyst for intramolecular C−N bond addition of amides to alkynes.
  • As a catalyst for cycloisomerization reactions of hydroxylated enynes.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Platinum- and Gold-Catalyzed Cycloisomerization Reactions of Hydroxylated Enynes
Victor Mamane, et al.
Journal of the American Chemical Society, 126, 8654-8655 (2004)
Intramolecular C?N Bond Addition of Amides to Alkynes Using Platinum Catalyst
Tomohiro Shimada, et al.
Journal of the American Chemical Society, 126, 10546-10547 (2004)
Solution-Assembled Ordered Grids Constructed with Silver Nanowires as Transparent Conductive Electrodes
De Li, et al.
ACS Omega, 3, 7191-7195 (2018)
Colloidal bimetallic platinum--ruthenium nanoparticles in ordered mesoporous carbon films as highly active electrocatalysts for the hydrogen evolution reaction
Rene Sachse, et al
Catalysis Science & Technology, 10, 2057-2068 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service