Skip to Content
Merck
All Photos(1)

Key Documents

520640

Sigma-Aldrich

Platinum(IV) chloride

≥99.9% trace metals basis

Synonym(s):

Platinum tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PtCl4
CAS Number:
Molecular Weight:
336.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Assay

≥99.9% trace metals basis

form

powder

composition

Pt, 55-58%

reaction suitability

reagent type: catalyst
core: platinum

impurities

≤1000.0 ppm Trace Metal Analysis

mp

370 °C (dec.) (lit.)

density

4.303 g/mL at 25 °C (lit.)

SMILES string

Cl[Pt](Cl)(Cl)Cl

InChI

1S/4ClH.Pt/h4*1H;/q;;;;+4/p-4

InChI key

FBEIPJNQGITEBL-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Platinum (IV)chloride is a weak Lewis acid used in surface modification, nanomaterialsynthesis, and catalysis.

Application

Platinum (IV) chloride can be used for the surfacemodification of crystalline and amorphous titania powders, to enhance theirphotocatalytic efficiency. Electronsinjected into the conduction band of titania due to visible light excitation ofplatinum (IV)chloride surface complex reduces oxygen to superoxide. The superoxide thusformed is responsible for the photodegradation reaction.

Platinumnanoparticles can be synthesized by reacting platinum (IV) chloride with Cyanobacteriumin an environmentally friendly method.

It can also be used as a dual catalyst forstereo- and regioselective glycosidations. Due to its ligand-bindingcapacity, platinum (IV) chloridehas an affinity for the glycosyl acceptor hydroxy group and forms the glycosidationproduct in β-configuration. It also enhances nucleophilicity differencesbetween hydroxy groups allowing regioselective glycosidation.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Macyk et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 7(9), 1862-1867 (2001-06-19)
Anatase, rutile, and amorphous titania powders were surface-modified by grinding with PtCl4 and H2[PtCl6]. Only the anatase modification afforded hybrid photocatalysts capable of degradation of 4-chlorophenol (4-CP) with visible light, with sufficient stability towards decomplexation. Grinding with K2[PtCl4] produced materials
Maggy F Lengke et al.
Langmuir : the ACS journal of surfaces and colloids, 22(17), 7318-7323 (2006-08-09)
Interaction of cyanobacteria (Plectonema boryanum UTEX 485) with aqueous platinum(IV)-chloride (PtCl(4) degrees ) has been investigated at 25-100 degrees C for up to 28 days, and 180 degrees C for 1 day. The addition of PtCl(4) degrees to the cyanobacteria
Barbora Nemcova et al.
Neuro endocrinology letters, 33 Suppl 3, 173-178 (2013-01-29)
Regarding the environmental pollution, platinum group elements (PGE) are in the centre of interest of current research. These rare elements are used as effective substances in automotive catalysts to reduce pollution by emissions originating from fuel combustion. Due to their
PtCl4-catalyzed domino synthesis of fused bicyclic acetals.
Alejandro Diéguez-Vázquez et al.
Angewandte Chemie (International ed. in English), 47(1), 209-212 (2007-12-07)
The mutagenic effect of platinum compounds in Drosophila melanogaster.
R C Woodruff et al.
Environmental mutagenesis, 2(2), 133-138 (1980-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service