Skip to Content
Merck
All Photos(2)

Key Documents

206091

Sigma-Aldrich

Platinum(II) chloride

98%

Synonym(s):

Platinum dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PtCl2
CAS Number:
Molecular Weight:
265.98
MDL number:
UNSPSC Code:
12161600
eCl@ss:
38191303
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

reaction suitability

core: platinum
reagent type: catalyst

mp

581 °C (dec.) (lit.)

density

6.05 g/mL at 25 °C

SMILES string

Cl[Pt]Cl

InChI

1S/2ClH.Pt/h2*1H;/q;;+2/p-2

InChI key

CLSUSRZJUQMOHH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Platinum (II) chloride is a catalyst used for a variety of C-C bond, C-O bond, and C-N bond formation.
Platinum (II) chloride can be used to catalyze the cyclization of 1‐(indole‐2‐yl)‐2,3‐allenols to form polysubstituted carbazoles. It is also used in studies involving metalation of aromatic compounds.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oyamada, J.; Kitamura, J.
Tetrahedron Letters, 46, 3823-3823 (2005)
Exclusive 1, 2-Aryl Shift in Platinum (II) Chloride-Catalyzed Cyclization of 1-(Indol-2-yl)-2, 3-allenols
Kong W, et al.
advanced synthesis and catalysis, 354(11-12), 2339-2347 (2012)
Yang, S. C.; Tsai, Y. C.; Shue, Y. J.
Organometallics, 20, 5326-5326 (2001)
Reactions of tri-tert-butylphosphine with platinum (II) and palladium (II). Facile intramolecular metalation of tri-tert-butylphosphine
Goel, Ram G and Montemayor, Reynaldo
Inorganic Chemistry, 16(9), 2183-2186 (1977)
Exclusive 1, 2-Aryl Shift in Platinum (II) Chloride-Catalyzed Cyclization of 1-(Indol-2-yl)-2, 3-allenols
Kong W, et al.
Advanced Synthesis & Catalysis, 354(11-12), 2339-2347 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service