Skip to Content
Merck
All Photos(3)

Documents

262587

Sigma-Aldrich

Chloroplatinic acid solution

8 wt. % in H2O

Synonym(s):

Hexachloroplatinic acid solution, Hydrogen hexachloroplatinate(IV) solution, Platinic chloride solution, Platinum(IV) chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2PtCl6
CAS Number:
Molecular Weight:
409.81
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

core: platinum
reagent type: catalyst

concentration

8 wt. % in H2O

density

1.05 g/mL at 25 °C

SMILES string

Cl.Cl.Cl[Pt](Cl)(Cl)Cl

InChI

1S/6ClH.Pt/h6*1H;/q;;;;;;+4/p-4

InChI key

GBFHNZZOZWQQPA-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Chloroplatinicacid (H2PtCl6) is a hygroscopic inorganic compound that is widely used as a catalyst for hydrosilylation, hydrogenation, carbonylation, oxidation and coupling reactions.

Chloroplatinic acid solution catalyses the hydrosilylation reaction of 1-Hexyne to form l,6-Bis(trichlorosilyl)hexane.

Application

Chloroplatinic acid solution used as:
  • Platinum precursor in the synthesis of platinum nanoparticles using hydrazine as a reducing agent.
  • An electrolyte in the preparation of various carbon-supported platinum nanomaterials with H2SO4 by electrodeposition method in the presence or absence of organic additives.
  • An anionic precursor in the preparation of Pt/SBA_BEA catalyst for the hydro isomerization of n-hexadecane.
  • A Pt precursor in the preparation of PVP-protected Pt-nanoparticles by seed-mediated growth method.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Composite of Pt/AlSBA-15+ zeolite catalyst for the hydroisomerization of n-hexadecane: The effect of platinum precursor
Fedyna M, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 305, 110366-110366 (2020)
The formation of 1, 6-bis (trichlorosilyl) hexane by the chloroplatinic acid catalyzed hydrosilylation of 1-hexyne
Benkeser RA, et al.
The Journal of Organic Chemistry, 32, 2634-2636 (1967)
Precursor Design for High Density Single Pt Atom Sites on MoS2: Enhanced Stability at Elevated Temperatures and Reduced 3D Clustering
Xiang Li, et al.
Chemistry of Materials, 32(6) , 2541?2551-2541?2551 (2020)
Synthesis of Platinum Particles from H2PtCl6 with Hydrazine as Reducing Agent
BEYRDBEY B, et al.
Gazi University Journal of Science, 22(4), 351-357 (2009)
B X E Desbiolles et al.
Nano letters, 20(6), 4520-4529 (2020-05-20)
Atomic force microscopy based approaches have led to remarkable advances in the field of mechanobiology. However, linking the mechanical cues to biological responses requires complementary techniques capable of recording these physiological characteristics. In this study, we present an instrument for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service