Skip to Content
Merck
All Photos(2)

Key Documents

520853

Sigma-Aldrich

Potassium tetrachloroplatinate(II)

≥99.9% trace metals basis

Synonym(s):

Potassium platinum(II) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2PtCl4
CAS Number:
Molecular Weight:
415.09
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder or crystals
Pricing and availability is not currently available.

grade

for analytical purposes

Assay

≥99.9% trace metals basis

form

powder or crystals

composition

Pt, ≥46.2%

impurities

≤1000.0 ppm Trace Metal Analysis

density

3.38 g/mL at 25 °C (lit.)

SMILES string

[K+].[K+].Cl[Pt--](Cl)(Cl)Cl

InChI

1S/4ClH.2K.Pt/h4*1H;;;/q;;;;2*+1;+2/p-4

InChI key

RVRDLMCWUILSAH-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Potassium tetrachloroplatinate(II) is a crystalline solid that is insoluble in water and soluble in organic solvents. It is widely used in the field of nanomaterial synthesis, catalysis, and energy storage.[1] [2]

Application

Potassium tetrachloroplatinate(II) can be used:
  • As a precursor to prepare platinum nanoparticles and mesoporous platinum nanospheres with tunable pore sizes.[1]
  • As a catalyst for selective methane oxidation through C-H activation. In addition, to synthesize octahedral [email protected] nanocrystals with an ultrathin PtNi alloy shell. The nanocrystals exhibited better ORR activity and durability than the commercial Pt/C for fuel cells.[2]
  • To prepare Pt atomic clusters were synthesized using a one-step carbon-defect-driven electroless deposition method. These catalysts demonstrated low overpotential, high activity for the hydrogen evolution reaction, exceptional stability, and improved utilization efficiency of Pt. The Pt-AC/DG (Atomic Clusters/Defective Graphene) cathode showed excellent hydrogen production activity, remarkable stability, and significantly reduced Pt usage compared to the Pt/C catalyst.[3]
  • As a starting material to synthesize cyclometalated platinum(II) complexes with long-lived luminescence in solution.[4]

Features and Benefits

Reacts with bis(dithiolates) to form metal-bis(dithiolates) with applications in laser Q-switch materials, liquid crystals, optical CD recording media, bar code materials and superconductivity. Reacts with capping polymers to form shape-control colloidal platinum nanoparticles for catalysis and photocatalysts.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alkyl chain functionalised, cyclometalated platinum(II) complexes: Syntheses, luminescence properties and X-ray crystal structure
Juliette A. Lowe, et al.
Journal of Organometallic Chemistry, 805, 87-93 (2016)
Selective Methane Oxidation Catalyzed by Platinum Salts in Oleum at Turnover Frequencies of Large-Scale Industrial Processes
Tobias Zimmermann, et al.
Journal of the American Chemical Society, 138, 12395-12400 (2016)
Polymeric micelle assembly for the smart synthesis of mesoporous platinum nanospheres with tunable pore sizes
Yunqi Li, et al.
Angewandte Chemie (International Edition in English), 54, 11073-11077 (2015)
Carbon-Defect-Driven Electroless Deposition of Pt Atomic Clusters for Highly Efficient Hydrogen Evolution
Qingqing Cheng, et al.
Journal of the American Chemical Society, 142, 5594-5601 (2020)
M Becker et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 133(1), 115-122 (1998-07-09)
The anticancer drug carboplatin has been monitored in rats during treatment by means of in vivo 195Pt NMR spectroscopy at 2.0 T. The purpose of the study was to assess local disposition kinetics in intact tissue following subcutaneous injection of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service