Skip to Content
Merck
All Photos(2)

Key Documents

206113

Sigma-Aldrich

Platinum(IV) chloride

96%

Synonym(s):

Platinum tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PtCl4
CAS Number:
Molecular Weight:
336.89
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38191307
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder and chunks

grade

for analytical purposes

Assay

96%

form

powder and chunks

reaction suitability

reagent type: catalyst
core: platinum

mp

370 °C (dec.) (lit.)

density

4.303 g/mL at 25 °C (lit.)

SMILES string

Cl[Pt](Cl)(Cl)Cl

InChI

1S/4ClH.Pt/h4*1H;/q;;;;+4/p-4

InChI key

FBEIPJNQGITEBL-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Platinum(IV) chloride can be used as a precursor to synthesize Pt-based nanocatalysts for various organic transformations. For example, Carbon-supported Pt-Ru nanoparticles can be used as an efficient catalyst for methanol and ethanol oxidation.

Due to its high affinity to the glycosyl acceptor hydroxy groups and bidentate ligation capacity, it can be used as a dual catalyst for regio- and stereoselective glycosidation reactions.

It can also be used for the photosensitization and surface modification of TiO2 for the photodegradation of 4-chlorophenol.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbora Nemcova et al.
Neuro endocrinology letters, 33 Suppl 3, 173-178 (2013-01-29)
Regarding the environmental pollution, platinum group elements (PGE) are in the centre of interest of current research. These rare elements are used as effective substances in automotive catalysts to reduce pollution by emissions originating from fuel combustion. Due to their
PtCl4-catalyzed domino synthesis of fused bicyclic acetals.
Alejandro Diéguez-Vázquez et al.
Angewandte Chemie (International ed. in English), 47(1), 209-212 (2007-12-07)
Ivana Bednarova et al.
Neuro endocrinology letters, 33 Suppl 3, 107-112 (2013-01-29)
An enhanced worldwide application of platinum group elements (PGE), in particular platinum, has been observed during recent decades. An increased concentration of PGE was determined in collected samples of great amount of aqueous ecosystems.The aim was to compare phytotoxic effect
N Kanematsu et al.
Gifu Shika Gakkai zasshi = The Journal of Gifu Dental Society, 17(2), 575-581 (1990-12-01)
The induction of mutation by cadmium chloride (CdCl2), platinum tetrachloride (PtCl4), and rhodium trichloride (RhCl3.3H2O) was measured utilizing the hypoxanthine-guanine phosphoribosyl transferase (HGPRT) locus in V-79 cells. Cadmium chloride at a concentration of 0.05 microM induced ford mutation in V-79
Lu Zhao et al.
Zhejiang da xue xue bao. Yi xue ban = Journal of Zhejiang University. Medical sciences, 38(1), 59-66 (2009-03-03)
To construct a drug carrier and gene vector PEG-PEI-Pt. Polyethyleneglycol (PEG) was coupled to polyethylenimine (PEI 600) and platinum tetrachloride; PEG-PEI-Pt complex was formed in ethanol. The complex was characterized by XRD, UV-VIS and FT-IR and the DNA condensation was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service