Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silver trifluoromethanesulfonate
Synonym(s):
Silver triflate, Trifluoromethanesulfonic acid silver salt, Silver trifluoromethanesulfonate, Ag(OTf), Silver (trifluoromethyl)sulfonate
Linear Formula:
CF
3
SO
3
Ag
CAS Number:
2923-28-6
Molecular Weight:
256.94
EC Number:
220-882-2
Products
Product Category
catalysts (1)
salts (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Special Grade
Purum (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
176435
Silver trifluoromethanesulfonate,
≥99%
Expand
View Pricing
483346
Silver trifluoromethanesulfonate,
≥99.95% trace metals basis
Expand
View Pricing
8.14325
Silver trifluoromethanesulfonate,
for synthesis
Expand
View Pricing
85325
Silver trifluoromethanesulfonate,
purum, ≥98.0% (Ag)
Expand
View Pricing