Skip to Content
Merck
All Photos(1)

Key Documents

176427

Sigma-Aldrich

Silver p-toluenesulfonate

≥99%

Synonym(s):

p-Toluenesulfonic acid silver salt, Silver tosylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO3Ag
CAS Number:
Molecular Weight:
279.06
Beilstein:
3598937
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Assay

≥99%

form

powder

reaction suitability

reagent type: catalyst
core: silver

SMILES string

[Ag+].Cc1ccc(cc1)S([O-])(=O)=O

InChI

1S/C7H8O3S.Ag/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);/q;+1/p-1

InChI key

JUDUFOKGIZUSFP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Converts alkyl halides to tosylates, and benzyl selenyl chlorides into their corresponding selenyl tosylates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 56, 2781-2781 (1991)
G Shanmugam et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 95, 177-183 (2012-05-29)
Bulk crystals of newly identified organic nonlinear optical material 4-methylanilinium p-toluenesulfonate (PMPT) were grown by slow evaporation solution growth method using ethanol as a solvent. It crystallizes in monoclinic system with a noncentrosymmetric space group P2(1). The formation of the
Mild C-H halogenation of anilides and the isolation of an unusual palladium(I)-palladium(II) species.
Robin B Bedford et al.
Angewandte Chemie (International ed. in English), 50(24), 5524-5527 (2011-05-14)
Hans H Adomat et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 902, 84-95 (2012-07-24)
Androgens are key mediators of prostate development and function, a role that extends to the development of prostate diseases such as benign prostatic hyperplasia (BPH) and prostate cancer. In prostate, DHT is the major androgen and reduction and glucuronidation are
Pei Liu et al.
Organic & biomolecular chemistry, 10(24), 4696-4698 (2012-05-18)
A convenient one-pot Mannich-type-cyclization-oxidation tandem process has been developed for the synthesis of 1,3,5-trisubstituted pyrazoles derivatives from aldehydes, hydrazines and alkynes using p-toluenesulfonic acid monohydrate (PTSA) as a multifunctional catalyst. This method provides a flexible and rapid route to 1,3,5-trisubstituted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service