Skip to Content
Merck
All Photos(2)

Documents

450804

Sigma-Aldrich

Copper(II) oxide

powder, 99.995% trace metals basis

Synonym(s):

Cupric oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuO
CAS Number:
Molecular Weight:
79.55
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99.995% trace metals basis

form

powder

application(s)

battery manufacturing

SMILES string

[Cu]=O

InChI

1S/Cu.O

InChI key

QPLDLSVMHZLSFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Janine Richter et al.
ChemistryOpen, 10(2), 97-109 (2021-02-11)
The application of ionic liquids for the dissolution of metal oxides is a promising field for the development of more energy- and resource-efficient metallurgical processes. Using such solutions for the production of valuable chemicals or electrochemical metal deposition requires a
Sumanta Kumar Meher et al.
Nanoscale, 5(5), 2089-2099 (2013-02-06)
In the quest to enhance the selectivity and sensitivity of novel structured metal oxides for electrochemical non-enzymatic sensing of glucose, we report here a green synthesis of unique sandwich-structured CuO on a large scale under microwave mediated homogeneous precipitation conditions.
Guangliang Cui et al.
Scientific reports, 3, 1250-1250 (2013-02-15)
Heterostructure material that acts as resonant tunneling system is a major scientific challenge in applied physics. Herein, we report a resonant tunneling system, quasi-2D Cu(2)O/SnO(2) p-n heterostructure multi-layer film, prepared by electrochemical deposition in a quasi-2D ultra-thin liquid layer. By
Hongwen Huang et al.
Nanoscale, 5(5), 1785-1788 (2013-01-31)
A facile, flexible and large-scale technique was proposed to prepare a CuO-CNT 3D-network composite with the aid of electrostatic interactions in aqueous solution. The composite greatly improves the electrochemical performance. At a rate of 0.1 C, the cycling discharge capacity
Wei Jian Foo et al.
Nanoscale, 5(2), 759-764 (2012-12-12)
Here we have demonstrated the preparation of high-quality, monodispersed and tunable phases of Cu nanoparticles. Structural and chemical composition studies depict the evolution of Cu-Cu(2)O-CuO nanoparticles at various process stages. The loading of Cu and Cu oxide nanoparticles on TiO(2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service