Merck
  • Non-noble metal Cu-loaded TiO2 for enhanced photocatalytic H2 production.
Non-noble metal Cu-loaded TiO2 for enhanced photocatalytic H2 production.
Nanoscale (2012-12-12)
Wei Jian Foo, Chun Zhang, Ghim Wei Ho
ABSTRACT
Here we have demonstrated the preparation of high-quality, monodispersed and tunable phases of Cu nanoparticles. Structural and chemical composition studies depict the evolution of Cu-Cu(2)O-CuO nanoparticles at various process stages. The loading of Cu and Cu oxide nanoparticles on TiO(2) catalyst has enhanced the photocatalytic H(2) production. Comparatively, H(2) treatment produces well-dispersed Cu nanoparticles with thin oxide shells that show the highest H(2) production amongst the samples. The relatively higher photocatalytic performance is deemed to result from reduced structural defects, higher surface area and dispersivity as well as favorable charge transfer, which inhibits recombination. The Cu nanoparticles are shown to be a promising alternative to noble metal-loaded TiO(2) catalyst systems due to their low cost and high performance in photocatalytic applications.
Sigma-Aldrich
Copper(II) oxide, powder, 99.995% trace metals basis
Sigma-Aldrich
Copper(II) oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Copper(II) oxide, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Copper(II) oxide, ACS reagent, ≥99.0%
Sigma-Aldrich
Copper(II) oxide, needles, mixture of CuO and Cu2O, ACS reagent
Sigma-Aldrich
Copper(II) oxide, powder, <10 μm, 98%