Skip to Content
Merck
All Photos(1)

Key Documents

774081

Sigma-Aldrich

Copper

nanopowder, 25 nm particle size (TEM)

Synonym(s):

Copper nanoparticles

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cu
CAS Number:
Molecular Weight:
63.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanopowder

resistivity

1.673 μΩ-cm, 20°C

particle size

25 nm (TEM)

bp

2567 °C (lit.)

mp

1083.4 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

SMILES string

[Cu]

InChI

1S/Cu

InChI key

RYGMFSIKBFXOCR-UHFFFAOYSA-N

General description

This copper nanopowder is a fine reddish-brown to black powder made of metallic copper nanoparticles with an average particle size of 25 nm. The particles are not oxidized and we recommend storing the product under inert gas to preserve the unoxidized surfaces.

Application

Copper nanopowder, 25 nm can be used for a variety of applications such as printed electronics, electroless copper plating, heat transfer fluids, catalysis, and thermal energy storage.

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Sol. 2 - Self-heat. 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chih-Ying Lee et al.
Chemosphere, 261, 128051-128051 (2020-10-30)
The potential toxicity of copper nanoparticles (CuNPs) to early stages of fishes is not fully understood, and little is known about their effects on ionocytes and associated functions. This study used zebrafish embryos as a model to investigate the toxic
Copper nanoparticles for printed electronics: routes towards achieving oxidation stability
Magdassi S, et al.
Materials, 3(9), 4626-4638 (2010)
Low-cost and high-throughput synthesis of copper nanopowder for nanofluid applications
Maji NC, et al.
Chemical Engineering Journal, 353, 34-45 (2018)
Evaluation of copper nanoparticles-Paraffin wax compositions for solar thermal energy storage
Lin SC and Al-Kayiem HH
Solar Energy, 132, 267-278 (2016)
Chan; G.H.;
Nano Letters, 7(7), 1947-1952 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service