Skip to Content
Merck
All Photos(1)

Documents

241741

Sigma-Aldrich

Copper(II) oxide

ACS reagent, ≥99.0%

Synonym(s):

Cupric oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuO
CAS Number:
Molecular Weight:
79.55
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Assay

≥99.0%

form

powder

reaction suitability

reagent type: catalyst
core: copper

impurities

Free alkali, passes test
≤0.002% N compounds
≤0.01% C compounds
≤0.02% S compounds
≤0.02% insol. dil. HCl

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.02%

cation traces

Ca: ≤0.01%
Fe: ≤0.05%
K: ≤0.02%
Na: ≤0.05%

SMILES string

[Cu]=O

InChI

1S/Cu.O

InChI key

QPLDLSVMHZLSFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Copper (II) oxide is one of the two stable oxides of copper. Cupric oxide is a precursor to many copper-containing products and chemicals. It is used as a pigment, fungicide, and antirust protection agent for marine paints. CuO possesses notable and admirable characteristics like abundance and affordability, ease of manufacturing, scalability, non-toxicity, antioxidant properties, and semiconducting characteristics.

Application

Copper (II) oxide is used:,As a catalyst in the synthesis of 2-Aryl- and 2-alkenyl-1,3,4-oxadiazoles from treating 1,3,4-oxadiazoles with aryl or alkenyl halides.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

DSC kinetics of the thermal decomposition of copper (II) oxalate by isoconversional and maximum rate (peak) methods.
Muraleedharan K and Kripa S.
Journal of Thermal Analysis and Calorimetry, 115(2), 1969-1978 (2014)
Copper (II) oxide nanowires for p-type conductometric NH3 sensing.
Shao F, et al.
Applied Surface Science, 311, 177-181 (2014)
Eagleson M.
Concise Encyclopedia Chemistry, 523-523 (1994)
A refinement of the crystal structure of copper (II) oxide with a discussion of some exceptional esd's.
?sbrink S and Norrby LJ.
Acta Crystallographica Section B, Structural Science, 26(1), 8-15 (1970)
Sumanta Kumar Meher et al.
Nanoscale, 5(5), 2089-2099 (2013-02-06)
In the quest to enhance the selectivity and sensitivity of novel structured metal oxides for electrochemical non-enzymatic sensing of glucose, we report here a green synthesis of unique sandwich-structured CuO on a large scale under microwave mediated homogeneous precipitation conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service