Skip to Content
Merck
All Photos(3)

Key Documents

203130

Sigma-Aldrich

Copper(II) oxide

99.999% trace metals basis

Synonym(s):

Cupric oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuO
CAS Number:
Molecular Weight:
79.55
EC Number:
MDL number:
UNSPSC Code:
12352303
eCl@ss:
38150108
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.999% trace metals basis

form

powder

application(s)

battery manufacturing

SMILES string

[Cu]=O

InChI

1S/Cu.O

InChI key

QPLDLSVMHZLSFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Auriane Fifame Oussou-Azo et al.
Nanomaterials (Basel, Switzerland), 10(5) (2020-05-28)
Copper has been used as an antimicrobial agent for over a century and is now being added to commercial fungicides. Nanomaterials have attracted much attention due to the special properties they have over their bulk form. We studied nanostructured copper
Pierre-Emmanuel Buffet et al.
Environmental science & technology, 47(3), 1620-1628 (2012-12-18)
The fate and effects of CuO nanoparticles (CuO NPs) were examined in endobenthic species (Scrobicularia plana , Hediste diversicolor), under environmentally realistic conditions in outdoor mesocosms (exposure to Cu at 10 μg L(-1) in particulate (CuO NPs) or soluble salt
Chengyi Hou et al.
Nanoscale, 5(3), 1227-1232 (2013-01-11)
We report a Cu(2)O nanocrystal-reduced graphene oxide hybrid that is dispersible in water and has anticancer activity under both visible and near-infrared light irradiation. In contrast to the highly efficient killing of both normal and cancer cells initiated by the
Sumanta Kumar Meher et al.
Nanoscale, 5(5), 2089-2099 (2013-02-06)
In the quest to enhance the selectivity and sensitivity of novel structured metal oxides for electrochemical non-enzymatic sensing of glucose, we report here a green synthesis of unique sandwich-structured CuO on a large scale under microwave mediated homogeneous precipitation conditions.
Lisha Zhou et al.
Nanoscale, 5(4), 1564-1569 (2013-01-18)
Stable Cu(2)O nanocrystals of around 3 nm were uniformly and densely grown on functionalized graphene sheets (FGS), which act as molecular templates instead of surfactants for controlled nucleation; the distribution density of nanocrystals can be easily controlled by FGS with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service