Skip to Content
Merck
All Photos(3)

Key Documents

203610

Sigma-Aldrich

Lead(II) oxide

99.999% trace metals basis

Synonym(s):

Lead monoxide, Plumbous oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbO
CAS Number:
Molecular Weight:
223.20
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

10 mmHg ( 0 °C)

Quality Level

Assay

99.999% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: lead

mp

886 °C (lit.)

SMILES string

O=[PbH2]

InChI

1S/O.Pb

InChI key

YEXPOXQUZXUXJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lead(II) oxide (PbO) is an industrially important metal oxide with strong absorption cross sections, high carrier mobilities, and conductivities. It is widely applied in the field of storage batteries, dielectrics, gas sensors, pigments, and paints.

Application

Lead(II) oxide can be used as a precursor to prepare:
  • Gamma ray radiation shielding glasses such as GeO2-PbO-Al2O3–CaO glasses.
  • Perovskite ceramic materials. For example, it can be used to synthesize lead magnesium niobate, widely applied in the field of electronic ceramics due to its high dielectric constant and voltage stability.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A - STOT RE 1

Target Organs

Central nervous system,Kidney,Blood

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Panizza et al.
Chemosphere, 90(4), 1455-1460 (2012-10-03)
In this paper the electrocatalytic properties of Ti-Ru-Sn ternary oxide (TiRuSnO(2)), PbO(2) and boron-doped diamond (BDD) anodes have been compared for the electrochemical oxidation of a real landfill leachate from an old municipal solid waste landfill (average values of COD
Guoting Li et al.
Journal of environmental sciences (China), 23(6), 998-1003 (2011-11-10)
Beta-PbO2 electrodes were prepared by electro-deposition and characterized by scanning electron microscopy, X-ray diffraction, X-ray photoelectron spectroscopy, and linear sweep voltammetry. We confirmed pure (beta-PbO2 crystals were on the electrode and it had a high oxygen evolution potential. The photoactivity
J Berashevich et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(7), 075803-075803 (2013-01-24)
The electronic properties of polycrystalline lead oxide consisting of a network of single-crystalline α-PbO platelets and the formation of native point defects in the α-PbO crystal lattice are studied using first-principles calculations. The results suggest that the polycrystalline nature of
Xingwen Lu et al.
Water research, 47(3), 1353-1360 (2013-01-05)
This study quantified the effects of lead stabilization by blending lead-containing sludge into glass-ceramics through sintering with aluminum- and silica-rich precursors. Using lead oxide (PbO) to simulate lead-laden sludge under thermal conditions, the predominant PbAl(2)O(4) phase was found at temperatures
E M Ibragimova et al.
Journal of nanoscience and nanotechnology, 12(11), 8818-8821 (2013-02-21)
Widely used SiO2-PbO glasses can be a good model for studying electron structure of insulator-metal interface with many Si-O-Pb bonds and eutectic Pb-Si mixture. Gamma-radiation of 60Co with an energy of -1.25 MeV generates efficiently electronic and hole structure defects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service