Skip to Content
Merck
All Photos(1)

Documents

449164

Sigma-Aldrich

Magnesium chloride

AnhydroBeads, −10 mesh, 99.9% trace metals basis

Synonym(s):

Magnogene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2
CAS Number:
Molecular Weight:
95.21
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

product line

AnhydroBeads

Quality Level

Assay

99.9% trace metals basis

impurities

≤1500.0 ppm Trace Metal Analysis

refractive index

n20/D 1.675 (lit.)

particle size

−10 mesh

mp

714 °C (lit.)

solubility

H2O: slightly soluble(lit.)

density

2.32 g/mL at 25 °C (lit.)

SMILES string

Cl[Mg]Cl

InChI

1S/2ClH.Mg/h2*1H;/q;;+2/p-2

InChI key

TWRXJAOTZQYOKJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium chloride (MgCl2) is an inorganic compound that can be prepared by reacting magnesium oxide and ammonium chloride in the presence of alumina as a covering agent.

Application

MgCl2 can be used in a variety of applications such as a precursor to magnesium metal, catalyst support in Ziegler-Natta catalyst and in food processing.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of anhydrous magnesium chloride from magnesia
Zhang Z, et al.
Industrial & Engineering Chemistry Research, 51(29), 9713-9718 (2012)
Magnesium compounds
Seeger M, et al.
Ullmann's Encyclopedia of Industrial Chemistry, 101(12), 4646-4655 (2000)
Commercially available metal alkyls and their use in polyolefin catalysts
Handbook of Transition Metal Polymerization Catalysts, 101(12), 1-28 (2010)
Pushkaraj R Patwardhan et al.
Bioresource technology, 101(12), 4646-4655 (2010-02-23)
Processing bio-oil with the help of currently existing petroleum refinery infrastructure has been considered as a promising alternative to produce sustainable fuels in the future. The feasibility of bio-oil production and upgrading processes depend upon its chemical composition which in
Elaine T Lim et al.
PLoS genetics, 10(7), e1004494-e1004494 (2014-08-01)
Exome sequencing studies in complex diseases are challenged by the allelic heterogeneity, large number and modest effect sizes of associated variants on disease risk and the presence of large numbers of neutral variants, even in phenotypically relevant genes. Isolated populations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service