Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Magnesium chloride
Synonym(s):
Magnogene, Magnesium chloride
Linear Formula:
MgCl
2
CAS Number:
7786-30-3
Molecular Weight:
95.21
EC Number:
232-094-6
Products
Product Category
salts (5)
Brand
Sigma-Aldrich (6)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Particle Size (µm)
Special Grade
Anhydrous (4)
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
M8266
Magnesium chloride,
anhydrous, ≥98%
Expand
View Pricing
208337
Magnesium chloride,
powder, <200 μm
Expand
View Pricing
8.14733
Magnesium chloride,
anhydrous for synthesis
Expand
View Pricing
M4880
Magnesium chloride,
BioReagent, suitable for insect cell culture, ≥97.0%
Expand
View Pricing
449164
Magnesium chloride,
AnhydroBeads
™
, −10 mesh, 99.9% trace metals basis
Expand
View Pricing
449172
Magnesium chloride,
AnhydroBeads
™
, −10 mesh, 99.99% trace metals basis
Expand
View Pricing