Skip to Content
Merck
All Photos(2)

Documents

339261

Sigma-Aldrich

Hydrofluoric acid

48 wt. % in H2O, ≥99.99% trace metals basis

Synonym(s):

HF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HF
CAS Number:
Molecular Weight:
20.01
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

vapor density

1.27 (vs air)
1.27 (vs air)

Quality Level

vapor pressure

25 mmHg ( 20 °C)

Assay

≥99.99% trace metals basis

form

liquid

concentration

48 wt. % in H2O

density

1.15 g/mL at 25 °C (lit.)

SMILES string

[H+].[F-]

InChI

1S/FH/h1H

InChI key

KRHYYFGTRYWZRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hydrofluoric acid (HF) is the 48% solution of HF in water. It is weakly acidic in nature. Aqueous HF is mostly used in etching silicon in semiconductors and producing inorganic fluoride compounds. HF can also be used as fluorinating and deprotecting agent. As fluorination agent it also forms complexes with organic bases.

Application

Hydrofluoric acid is used:
  • To compose the electrolyte for the synthesis of TiO2 nanotube arrays.
  • As etchant for the glass fibers.
  • As capping reagent for the synthesis of anatase TiO2 nanocrystals/nanosheets.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly-ordered TiO2 nanotube arrays up to 220μm in length: use in water photoelectrolysis and dye-sensitized solar cells.
Shankar K, et al.
Nanotechnology, 18(6), 065707-065707 (2007)
Francesca Monticelli et al.
Dental materials : official publication of the Academy of Dental Materials, 22(7), 602-609 (2005-11-18)
To verify the influence of different etching procedures of the post-surface on microtensile bond strength values between fiber posts and composite core materials. 60 DT Light Posts were divided into 10 subgroups using five different chemical surface treatments and two
Eagleson M.
Concise Encyclopedia Chemistry, 508-508 (1994)
Anatase TiO2 nanosheet: an ideal host structure for fast and efficient lithium insertion/extraction.
Chen JS and Lou XW.
Electrochemical Communications, 11(12), 2332-2335 (2009)
Nicolas Vodovar et al.
JACC. Heart failure, 3(8), 629-636 (2015-08-08)
This study sought to hypothesize that elevated B-type natriuretic peptide (BNP) could act as an endogenous neprilysin inhibitor. A hallmark of acute decompensated heart failure (ADHF) is the overproduction of natriuretic peptides (NPs) by stretched cardiomyocytes. Various strategies have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service