Skip to Content
Merck
All Photos(1)

Key Documents

449172

Sigma-Aldrich

Magnesium chloride

AnhydroBeads, −10 mesh, 99.99% trace metals basis

Synonym(s):

Magnogene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2
CAS Number:
Molecular Weight:
95.21
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis

product line

AnhydroBeads

Quality Level

Assay

99.99% trace metals basis

impurities

≤150.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

714 °C (lit.)

density

2.32 g/mL at 25 °C (lit.)

SMILES string

Cl[Mg]Cl

InChI

1S/2ClH.Mg/h2*1H;/q;;+2/p-2

InChI key

TWRXJAOTZQYOKJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium chloride can be used as a starting material to prepare electrolyte solutions for Mg-polysulfide flow batteries.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of chloride for a simple, non-grignard Mg electrolyte in ether-based solvents
Niya Sa, et al
ACS Applied Materials & Interfaces, 8, 16002-16008 (2016)
Stability and Disproportionation of Magnesium Polysulfides and the Effects on the Mg-Polysulfide Flow Battery
Peng He, et al.
Journal of the Electrochemical Society, 168, 110516-110516 (2021)
Kadri Õunap et al.
PloS one, 10(7), e0133841-e0133841 (2015-07-28)
The human WBSCR22 protein is a 18S rRNA methyltransferase involved in pre-rRNA processing and ribosome 40S subunit biogenesis. Recent studies have shown that the protein function in ribosome synthesis is independent of its enzymatic activity. In this work, we have
Manal A Farg et al.
Human molecular genetics, 23(13), 3579-3595 (2014-02-20)
Intronic expansion of a hexanucleotide GGGGCC repeat in the chromosome 9 open reading frame 72 (C9ORF72) gene is the major cause of familial amyotrophic lateral sclerosis (ALS) and frontotemporal dementia. However, the cellular function of the C9ORF72 protein remains unknown.
Takeshi Tomita et al.
PloS one, 9(9), e108957-e108957 (2014-10-01)
Celastramycin A, a small molecule that inhibits the production of antibacterial peptides in an ex vivo culture system of Drosophila, suppresses the TNFα-mediated induction of IL-8 in mammalian cells. To understand its molecular mechanism, we examined Celastramycin A binding proteins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service