Skip to Content
Merck
All Photos(1)

Key Documents

499609

Sigma-Aldrich

Calcium chloride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Calcium dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCl2
CAS Number:
Molecular Weight:
110.98
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
grade:
anhydrous
form:
powder
solubility:
ethanol: very soluble(lit.)
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor pressure

0.01 mmHg ( 20 °C)

Assay

99.99% trace metals basis

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

≤150.0 ppm Trace Metal Analysis

bp

1935 °C/1 atm (lit.)

mp

772 °C (lit.)

solubility

ethanol: very soluble(lit.)

SMILES string

[Cl-].[Cl-].[Ca++]

InChI

1S/Ca.2ClH/h;2*1H/q+2;;/p-2

InChI key

UXVMQQNJUSDDNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium chloride is an inorganic compound widely used in desiccators, catalysis, and thermal storage applications. It has the following properties:
  • Higher thermal conductivity
  • Chemical and thermal stability
  • Non-toxic and less corrosive

Application

Calcium chloride can be used:
  • As electrolyte in the reduction of solid zirconium dioxide by electro-deoxidation.
  • A raw material to prepare composite adsorbent(activated carbon/silica gel/CaCl2) for cooling and dehumidification system.
  • As a catalyst to synthesize tetrahydropyranyl ethers via tetrahydropyranylation of alcohols and phenols.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Levi et al.
Journal of thrombosis and haemostasis : JTH, 12(9), 1428-1436 (2014-05-09)
Four-factor prothrombin complex concentrates (PCCs), which contain factor II, FVII, FIX, and FX, have shown the potential to reverse the anticoagulant effect of rivaroxaban in healthy volunteers. The purpose of this study was to determine whether a three-factor PCC, which contains
J Philippe et al.
International journal of obesity (2005), 39(2), 295-302 (2014-06-04)
A significant proportion of severe familial forms of obesity remain genetically elusive. Taking advantage of our unique cohort of multigenerational obese families, we aimed to assess the contribution of rare mutations in 29 common obesity-associated genes to familial obesity, and
Maheswari Jayakannan et al.
Journal of experimental botany, 66(7), 1865-1875 (2015-01-24)
The role of endogenous salicylic acid (SA) signalling cascades in plant responses to salt and oxidative stresses is unclear. Arabidopsis SA signalling mutants, namely npr1-5 (non-expresser of pathogenesis related gene1), which lacks NPR1-dependent SA signalling, and nudt7 (nudix hydrolase7), which
Meral Tunc-Ozdemir et al.
Plant physiology, 161(2), 1010-1020 (2013-02-02)
Cyclic nucleotide-gated channels (CNGCs) have been implicated in diverse aspects of plant growth and development, including responses to biotic and abiotic stress, as well as pollen tube growth and fertility. Here, genetic evidence identifies CNGC16 in Arabidopsis (Arabidopsis thaliana) as
Mohsen Akbari et al.
Advanced functional materials, 24(26), 4060-4067 (2014-11-21)
The fabrication of cell-laden structures with anisotropic mechanical properties while having a precise control over the distribution of different cell types within the constructs is important for many tissue engineering applications. Automated textile technologies for making fabrics allow simultaneous control

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Does anyone know how to open the glass ampoule that it comes in? It's just one long tube with no neck.

    1 answer
    1. In order to open the glass ampoule, it is recommended to use an ampoule opener such as Product Z693847 (Scienceware® Break-Safe ™ ampule opener). Here is a short video on how to use the product:
      https://www.sigmaaldrich.com/video/labware/bel-art-ampule-opener.html

      An alternative solution is to groove the bulb using a glass cutting tool such as the following:
      - Ref. Z169064: diamond glass cutter
      - Ref. Z150770: glass tube cutter

      After marking the circumference of the bulb evenly, grab puncture resistant gloves or nitrile gloves and a thick cloth to wrap around the bulb. The bulk can then be broken by exerting pressure on the marking with the end-user's thumb.

      Note that the ampoules are intended for single use. After the bulb is broken, it is suggested to use the product at one time. It is not recommended to store unused surplus. To keep the unused product after opening, it is recommended to transfer the unused material to another clean, properly sealed container for storage.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service