Skip to Content
Merck
All Photos(1)

Key Documents

262781

Sigma-Aldrich

Niobium

foil, thickness 0.25 mm, 99.8% trace metals basis

Synonym(s):

Columbium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Nb
CAS Number:
Molecular Weight:
92.91
EC Number:
MDL number:
UNSPSC Code:
12141730
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.8% trace metals basis

form

foil

resistivity

13-16 μΩ-cm, 20°C

thickness

0.25 mm

bp

4742 °C (lit.)

mp

2468 °C (lit.)

density

8.57 g/mL at 25 °C (lit.)

SMILES string

[Nb]

InChI

1S/Nb

InChI key

GUCVJGMIXFAOAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Quantity

10.7 g = 50 × 100 mm; 32.1 g = 100 × 150 mm

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M I Tsindlekht et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(8), 085701-085701 (2013-01-31)
We report results of measurements of the ac conductivity of a Nb superconducting thin film in a swept dc magnetic field. In the mixed state the swept dc field creates vortices at the film surface which pass through the film
Huaxiang Yi et al.
Optics express, 20(25), 27562-27568 (2012-12-25)
We demonstrate error-free 80km transmission by a silicon carrier-depletion Mach-Zehnder modulator at 10Gbps and the power penalty is as low as 1.15dB. The devices were evaluated through the bit-error-rate characterizations under the system-level analysis. The silicon Mach-Zehnder modulator was also
Evgeniy Papulovskiy et al.
Physical chemistry chemical physics : PCCP, 15(14), 5115-5131 (2013-03-02)
Ab initio DFT calculations of (93)Nb NMR parameters using the NMR-CASTEP code were performed for a series of over fifty individual niobates, and a good agreement has been found with experimental NMR parameters. New experimental and calculated (93)Nb NMR data
Yoon-Jun Kim et al.
ACS nano, 7(1), 732-739 (2012-12-25)
Imaging the three-dimensional atomic-scale structure of complex interfaces has been the goal of many recent studies, due to its importance to technologically relevant areas. Combining atom-probe tomography and aberration-corrected scanning transmission electron microscopy (STEM), we present an atomic-scale study of
Anjan Soumyanarayanan et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(5), 1623-1627 (2013-01-16)
The competition between proximate electronic phases produces a complex phenomenology in strongly correlated systems. In particular, fluctuations associated with periodic charge or spin modulations, known as density waves, may lead to exotic superconductivity in several correlated materials. However, density waves

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service