Merck
  • Demonstration of low power penalty of silicon Mach-Zehnder modulator in long-haul transmission.
Demonstration of low power penalty of silicon Mach-Zehnder modulator in long-haul transmission.
Optics express (2012-12-25)
Huaxiang Yi, Qifeng Long, Wei Tan, Li Li, Xingjun Wang, Zhiping Zhou
ABSTRACT
We demonstrate error-free 80km transmission by a silicon carrier-depletion Mach-Zehnder modulator at 10Gbps and the power penalty is as low as 1.15dB. The devices were evaluated through the bit-error-rate characterizations under the system-level analysis. The silicon Mach-Zehnder modulator was also analyzed comparatively with a lithium niobate Mach-Zehnder modulator in back-to-back transmission and long-haul transmission, respectively, and verified the negative chirp parameter of the silicon modulator through the experiment. The result of low power penalty indicates a practical application for the silicon modulator in the middle- or long-distance transmission systems.
Sigma-Aldrich
Lithium niobate, 99.9% trace metals basis
Niobium, IRMM®, certified reference material, 0.5 mm wire
Niobium, IRMM®, certified reference material, 0.5 mm wire
Niobium, IRMM®, certified reference material, 0.02 mm foil
Sigma-Aldrich
Niobium, powder, <45 μm, 99.8% trace metals basis
Sigma-Aldrich
Niobium, foil, thickness 0.25 mm, 99.8% trace metals basis
Niobium, IRMM®, certified reference material, 0.02 mm foil