Skip to Content
Merck
All Photos(2)

Key Documents

262722

Sigma-Aldrich

Niobium

powder, <45 μm, 99.8% trace metals basis

Synonym(s):

Columbium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Nb
CAS Number:
Molecular Weight:
92.91
EC Number:
MDL number:
UNSPSC Code:
12141730
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.8% trace metals basis

form

powder

resistivity

13-16 μΩ-cm, 20°C

particle size

<45 μm

bp

4742 °C (lit.)

mp

2468 °C (lit.)

density

8.57 g/mL at 25 °C (lit.)

SMILES string

[Nb]

InChI

1S/Nb

InChI key

GUCVJGMIXFAOAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M I Tsindlekht et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(8), 085701-085701 (2013-01-31)
We report results of measurements of the ac conductivity of a Nb superconducting thin film in a swept dc magnetic field. In the mixed state the swept dc field creates vortices at the film surface which pass through the film
Huaxiang Yi et al.
Optics express, 20(25), 27562-27568 (2012-12-25)
We demonstrate error-free 80km transmission by a silicon carrier-depletion Mach-Zehnder modulator at 10Gbps and the power penalty is as low as 1.15dB. The devices were evaluated through the bit-error-rate characterizations under the system-level analysis. The silicon Mach-Zehnder modulator was also
Christos D Malliakas et al.
Journal of the American Chemical Society, 135(5), 1719-1722 (2013-01-23)
2H-NbSe(2) is a canonical Charge-Density-Wave (CDW) layered material the structural details of which remained elusive. We report the detailed structure of 2H-NbSe(2) below the CDW transition using a (3 + 2)-dimensional crystallographic approach on single crystal X-ray diffraction data collected
Evgeniy Papulovskiy et al.
Physical chemistry chemical physics : PCCP, 15(14), 5115-5131 (2013-03-02)
Ab initio DFT calculations of (93)Nb NMR parameters using the NMR-CASTEP code were performed for a series of over fifty individual niobates, and a good agreement has been found with experimental NMR parameters. New experimental and calculated (93)Nb NMR data
Tzyy-Jiann Wang et al.
Optics express, 20(27), 28119-28124 (2012-12-25)
We report ultra-smooth LiNbO(3) microdisk resonators fabricated by selective ion implantation, chemical etching, and thermal treatment. The undercut microdisk structure is produced by chemically etching the buried lattice damage layer formed by selective ion implantation. By thermal treatment, surface tension

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service