Skip to Content
Merck
All Photos(1)

Documents

357162

Sigma-Aldrich

Vanadium

foil, thickness 0.127 mm, 99.7% trace metals basis

Synonym(s):

Atomic vanadium, Vanadium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
V
CAS Number:
Molecular Weight:
50.94
EC Number:
MDL number:
UNSPSC Code:
12141749
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

8 mmHg ( 20 °C)

Assay

99.7% trace metals basis

form

foil

resistivity

24.8-26.0 μΩ-cm, 20°C

thickness

0.127 mm

bp

3380 °C (lit.)

mp

1890 °C (lit.)

density

6.11 g/mL at 25 °C (lit.)

SMILES string

[V]

InChI

1S/V

InChI key

LEONUFNNVUYDNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Vanadium foil can be used for a variety of applications such as:
  • preparation of vanadium pentoxide for lithium ion batteries
  • starting material for synthesis of vanadium oxide
  • a substrate for the deposition of aluminium oxide for wear resistant coatings

Quantity

1.9 g = 50 × 50 mm; 7.6 g = 100 × 100 mm

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanadium pentoxide cathode materials for high-performance lithium-ion batteries enabled by a hierarchical nanoflower structure via an electrochemical process
Tang Y, et al.
Journal of Material Chemistry A, 1(1), 82-88 (2013)
XPS study of vanadium surface oxidation by oxygen ion bombardment
Alov N, et al.
Surface Science, 600(8), 1628-1631 (2006)
Characterization of alpha-phase aluminum oxide films deposited by filtered vacuum arc
Yamada-Takamura Y, et al.
Surface and Coatings Technology, 142(8), 260-264 (2001)
Energy storage studies of bare and doped vanadium pentoxide,(V 1.95 M 0.05) O 5, M= Nb, Ta, for lithium ion batteries
Sakunthala A, et al.
Energy & Environmental Science, 4(5), 1712-1725 (2011)
Nolan S Horner et al.
Analytica chimica acta, 758, 28-35 (2012-12-19)
A previously developed method based on continuous on-line leaching with artificial gastro-intestinal fluids was used to determine the bio-accessible fraction of As, Cu, Fe, V and Zn in brown and white rice from California by inductively coupled mass spectrometry (ICP-MS).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service