Skip to Content
Merck
All Photos(4)

Key Documents

203920

Sigma-Aldrich

Niobium(V) oxide

99.99% trace metals basis

Synonym(s):

Niobium pentoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Nb2O5
CAS Number:
Molecular Weight:
265.81
EC Number:
MDL number:
UNSPSC Code:
12352303
eCl@ss:
38180502
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.99% trace metals basis

form

powder

density

4.47 g/mL at 25 °C (lit.)

SMILES string

O=[Nb](=O)O[Nb](=O)=O

InChI

1S/2Nb.5O

InChI key

ZKATWMILCYLAPD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Niobium(V) oxide, also known as niobium pentoxide, is a white crystalline powder. It is a refractory metal oxide with a high melting point of over 2500 °C and high chemical stability, including to oxygen and moisture. Niobium(V) oxide is mainly used as a raw material in the production of niobium and niobium alloys, as a catalyst in chemical reactions, and as a refractory material in high-temperature applications. It is also used as a dielectric material in capacitors and as a pigment in ceramics and glasses.

Application

Applied in the solid state formation of an unusual cation deficient perovskite, Ba7Nb4MoO20.
Promising results were obtained using niobium(V) oxide as an alternate electrode to lithium metal in advanced fuel cells.

Features and Benefits

Applied in the solid state formation of an unusual cation deficient perovskite, Ba7Nb4MoO20.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Garcia-Gonzalez, E. et al.
Chemistry of Materials, 11, 433-433 (1999)
Hong-Yi Lin et al.
Journal of biomedical nanotechnology, 5(2), 215-223 (2010-01-09)
The feasibility of using niobium oxide-coated magnetic nanoparticles (NPs) as affinity probes for selectively trapping phosphopeptides from peptide mixtures including tryptic digest of caseins, serum, and cell lysate was demonstrated in this study. Phosphopeptide enrichment was rapid when subjecting these
Jinwoo Lee et al.
Nature materials, 7(3), 222-228 (2008-01-29)
Even after a decade or so of research, the direct synthesis of highly crystalline mesoporous transition-metal oxides that are thermally stable and well ordered still constitutes a major challenge. Although various soft- and hard-templating approaches have been developed in the
Takashi Hisatomi et al.
Advanced materials (Deerfield Beach, Fla.), 24(20), 2699-2702 (2012-04-18)
A 2-nm thick Nb(2)O(5) underlayer deposited by atomic layer deposition increases the charge separation efficiency and the photovoltage of ultrathin hematite films by suppressing electron back injection. Absorbed photon-to-current efficiencies (APCE) as high as 40%, which are one of the
Jian-Lang Li et al.
Optics express, 16(14), 10841-10848 (2008-07-09)
Cylindrical vector beams were produced from laser diode end-pumped Nd:YAG ceramic microchip laser by use of two types of subwavelength multilayer gratings as the axisymmetric-polarization output couplers respectively. The grating mirrors are composed of high- and low-refractive- index (Nb(2)O(5)/SiO(2)) layers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service