Skip to Content
Merck
All Photos(3)

Key Documents

334472

Sigma-Aldrich

Hydrogen hexahydroxyplatinate(IV)

99.9% trace metals basis

Synonym(s):

Platinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2Pt(OH)6
CAS Number:
Molecular Weight:
299.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder and chunks
solubility:
H2O: slightly soluble(lit.)
aqueous acid: slightly soluble(lit.)
Pricing and availability is not currently available.

grade

for analytical purposes

Assay

99.9% trace metals basis

form

powder and chunks

impurities

≤1500.0 ppm Trace Metal Analysis

solubility

H2O: slightly soluble(lit.)
aqueous acid: slightly soluble(lit.)

SMILES string

O.O.O[Pt](O)(O)O

InChI

1S/6H2O.Pt/h6*1H2;/q;;;;;;+4/p-4

InChI key

ILXDKMTYQJUXAB-UHFFFAOYSA-J

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianjian Lin et al.
Scientific reports, 4, 5769-5769 (2014-08-30)
Three-dimensional (3D) hierarchical nanoscale architectures comprised of building blocks, with specifically engineered morphologies, are expected to play important roles in the fabrication of 'next generation' microelectronic and optoelectronic devices due to their high surface-to-volume ratio as well as opto-electronic properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service