Skip to Content
Merck
All Photos(3)

Key Documents

254770

Sigma-Aldrich

Potassium ethyl xanthogenate

96%

Synonym(s):

O-Ethylxanthic acid potassium salt, KEX, Potassium O-ethyl dithiocarbonate, Potassium xanthogenate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCSSK
CAS Number:
Molecular Weight:
160.30
Beilstein:
3596974
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

powder

mp

210 °C (dec.) (lit.)

solubility

alcohol: soluble(lit.)
water: very soluble(lit.)

SMILES string

[K+].CCOC([S-])=S

InChI

1S/C3H6OS2.K/c1-2-4-3(5)6;/h2H2,1H3,(H,5,6);/q;+1/p-1

InChI key

JCBJVAJGLKENNC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Potassium ethyl xanthogenate (ethyl potassium xanthogenate) has been used:
  • in synthesis of unsymmetrical sulfides via cross-coupling reaction, using recyclable CuO nanoparticles under ligand-free conditions
  • as surfactant to investigate the adsorption of colloidal dye Disperse Blue 3 onto pretreated polyester fabric
  • in determination of microgram amounts of cadmium in water samples by substoichiometric radiochemical method

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

204.8 °F

Flash Point(C)

96 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Tjälve et al.
The Science of the total environment, 148(2-3), 217-242 (1994-06-06)
Dithiocarbamates, thiuram sulphides, xanthates, pyridinethiones and halogenated 8-hydroxyquinolines are groups of compounds which can form lipophilic complexes with Ni2+. These compounds are widely used as drugs and pesticides, and in industry. We have exposed rodents (mice, rats) and fish (brown
Anthony J Shallop et al.
The Journal of organic chemistry, 68(22), 8657-8661 (2003-10-25)
We have previously reported the use of a 13C tag at the C2 of 15N-multilabeled purine nucleosides to distinguish the adjacent-labeled 15N atoms from those in an untagged nucleoside. We now introduce the use of an indirect tag at the
M C De Ungria et al.
Helicobacter, 3(4), 269-277 (1998-12-09)
Plasmids are extra-chromosomal DNA that may encode products that aid in virulence, pathogenesis, and the spread of antibiotic resistance among a wide spectrum of bacteria. Plasmids have been detected in Helicobacter pylori, H. felis, H. fennelliae, and H. cinaedi. However
Yi-Kuo Chang et al.
Journal of hazardous materials, 94(1), 89-99 (2002-07-27)
Although, the xanthate process has been shown to be an effective method for heavy metal removal from contaminated water, a heavy metal contaminated residual sludge is produced by the treatment process and the metal-xanthate sludge must be handled in accordance
M Espinosa-Jiménez et al.
Journal of colloid and interface science, 265(2), 227-233 (2003-09-10)
Data are presented on the adsorption of the colloidal dye Disperse Blue 3 onto polyester fabric (Dacron 54, Stile 777), the fabric being pretreated with different amounts of the surfactant potassium ethyl xanthogenate (PEX). This study has been made by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service