Merck
GC Analysis of FAMEs in Fish Oil Capsules on Omegawax
GC Analysis of FAMEs in Fish Oil Capsules on Omegawax suitable for GC
材料
分析柱
產品號碼
描述
訂價
Omegawax 毛细管GC色谱柱
L × I.D. 30 m × 0.25 mm, df 0.25 μm
相關產品
產品號碼
描述
訂價
三氟化硼-甲醇
10 % (w/w), pkg of 20 × 1 mL
三氟化硼-甲醇
10 % (w/w)
CONDITIONS
column
Omegawax, 30 m x 0.25 mm I.D., 0.25μm (24136)
oven
170 °C, 1 °C/min to 225°C
inj. temp.
250 °C
detector
FID 260 °C
carrier gas
helium, 1.2 mL/min
injection
1 μL, 100:1 split
liner
4 mm ID FocusLiner with taper
sample
1. Fish Oil from capsule, prepared according to AOAC Method 991.39 2. Atlantic, farm-raised salmon, prepared according to AOCS Official Method Ce 1k-09
描述
分析報告
Samples of a commercially purchased fish oil capsule were analyzed for fatty acids. Sample preparation of the fish oil capsule was done as described in AOAC Official Method 991.39 (1). The resulting extracts were then analyzed by GC/FID on an Omegawax 250 capillary column as described in AOCS Official Method Ce 1i-07.