跳轉至內容
Merck
全部照片(1)

重要文件

5.04540

Sigma-Aldrich

EBP1 Inhibitor, WS6

同義詞:

EBP1 Inhibitor, WS6, N-(6-(4-(2-(4-((4-Methylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenylamino)-2-oxoethyl)phenoxy)pyrimidin-4-yl)cyclopropanecarboxamide, xHCl, Erb3 Binding Protein-1 Inhibitor, IKK Inhibitor XV, PA2G4 Inhibitor

登入查看組織和合約定價


About This Item

經驗公式(希爾表示法):
C29H31F3N6O3 · xHCl
分子量::
568.59 (free base basis)
分類程式碼代碼:
12352200
PubChem物質ID:
暫時無法取得訂價和供貨情況

化驗

≥98% (HPLC)

品質等級

形狀

powder

製造商/商標名

Calbiochem®

儲存條件

OK to freeze
desiccated (hygroscopic)
protect from light

顏色

white

溶解度

DMSO: 100 mg/mL
water: 100 mg/mL

儲存溫度

−20°C

SMILES 字串

CN1CCN(CC1)CC2=C(C=C(C=C2)NC(=O)CC3=CC=C(C=C3)OC4=NC=NC(=C4)NC(=O)C5CC5)C(F)(F)F

一般說明

A cell-permeable phenoxypyrimidin-cyclopropanecarboxamide compound that effectively stimulates the growth of primary β cells in cultures in vitro (ECmax in 4 d = 200 and 400 nM, respectively, for increasing Ki-67-positive β cell population in primary rat and human islets) and in the murine type 1 diabetes (T1D) model RIP-DTA in vivo (170% of control pancreas Ki-67-positive β cells in 1 wk and 150% of control pancreas β cell mass in 6 wks; 5 mg/kg i.p. q.o.d.) with significant blood glucose reducing efficacy (200 vs. 425 mg/dL on d45 with or without treatment). WS6 is shown to interact with IKKε and EBP1/PA2G4/Erb3 binding protein-1 as well as upregulate cellular E2F mRNA (2.8- and 4.8-fold of vehicle control, respectively, with 8 h and 12 h treatment; 1 µM) and transcription activity (1.4- and 1.5-fold of vehicle control, respectively, at 0.74 and 2.22 µM; 24 h treatment) in murine β cell line R7T1, consistent with blocking the known suppressor function of EBP1 toward E2F-mediated transcription. Likewise, dual inhibition of EBP1 (with shRNA) and IKK (1 µM; Cat. Nos. 401486 & 505378) is shown to fully recapitulate the proliferating effect of WS6 in R7T1 cells.

生化/生理作用

Cell permeable: yes
Primary Target
EBP1/PA2G4/Erb3 binding protein-1
Reversible: yes
Secondary Target
IKKε

包裝

Packaged under inert gas

警告

Toxicity: Standard Handling (A)

重構

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

其他說明

Shen, W., et al. 2013. J. Am. Chem. Soc.135, 1669.

法律資訊

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

儲存類別代碼

11 - Combustible Solids

水污染物質分類(WGK)

WGK 3

閃點(°F)

Not applicable

閃點(°C)

Not applicable


分析證明 (COA)

輸入產品批次/批號來搜索 分析證明 (COA)。在產品’s標籤上找到批次和批號,寫有 ‘Lot’或‘Batch’.。

已經擁有該產品?

您可以在文件庫中找到最近購買的產品相關文件。

存取文件庫

Questions

Reviews

No rating value

Active Filters

我們的科學家團隊在所有研究領域都有豐富的經驗,包括生命科學、材料科學、化學合成、色譜、分析等.

聯絡技術服務