SU860065
1 of 10
Supelco
27293
顶空样品瓶(透明玻璃材质,斜顶平底)
30228-U
Supel™惰性多层箔
30226-U
PHL89262
槲皮素
27294
斜顶圆底顶空样品瓶
Z272078
NMR tube caps
27455-U
带 PTFE/硅胶隔垫的钳口密封盖
PHL83535
Quercetin 3-O-rutinoside-7-O-glucoside
PHR1488
30227-U
beveled finish clear glass round bottom
closure type crimp top vial
pkg of 100 ea
22 mm × 38.2 mm × 12.5 mm
5 mL
for crimp seal
正在寻找类似产品? 访问 产品对比指南
pressure release open center
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.