Kluczowe dokumenty
58462
Supelco
Dynaseal Fittings
Set of nuts, collet, and ferrule
About This Item
Kod UNSPSC:
41115700
opakowanie
pkg of 2 ea
dopasowywanie
10-32 male Dynaseal nut and ferrule for 1/16 in. O.D. tubing (collet included)
Cechy i korzyści
Dynaseal hand-tight fittings will seal against pressures to over 7000psi/490kg/cm². Each fitting consists of a reusable nut, collet, and ferrule. The polymer ferrule will not deform stainless steel seats and can be reused. Except for the fitting for Rheodyne valves, these fittings are compatible with all 1/16 in. female compression-type fittings now used on HPLC columns.
Polecane produkty
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Informacje prawne
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)