The SPE Vacuum Manifold does not include a pump but can be used with any vacuum pump available in most laboratories. This vacuum manifold is compatible with any SPE tubes that have a standard luer lock connection.
Przejdź do
Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 9360,00 zł |
Informacje o tej pozycji
9360,00 zł
Quality Segment
material
PTFE frit, polypropylene tube
product line
Visiprep™
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
General description
Application
Features and Benefits
- Patented screw-type valves within each SPE port for precise flow control
- Glass basin will not dissolve, fog, or discolor when exposed to solvents
- Legs on cover allow user to easily rest cover on work surface when removed from the manifold
- Screw-type solvent-resistant vacuum bleed gauge and valve offer better sealing and vacuum control. Valve takes
1/4 ″ vacuum tubing. - PP collection vessel rack accommodates autosampler vials; small scintillation vials (22.75 mm O.D. recommended); 10 and 16 mm test tubes; and 1, 2, 5, and 10 mL volumetric flasks.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description DL (Disposable Liner), 24-port model | description DL (Disposable Liner), 12-port model | description for use with DL (Disposable Liner) 12- and 24-port models, pk of 2 | description for use with standard 12- and 24-port models, pk of 2 |
| application(s) food and beverages | application(s) environmental | application(s) food and beverages | application(s) food and beverages |
| technique(s) solid phase extraction (SPE): suitable | technique(s) solid phase extraction (SPE): suitable | technique(s) solid phase extraction (SPE): suitable | technique(s) solid phase extraction (SPE): suitable |
| material polypropylene tube, PTFE frit | material polypropylene tube | material - | material - |
| product line Visiprep™ | product line - | product line Visiprep™ | product line Visiprep™ |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
Still not finding the right product?
Explore all of our products under Visiprep™ SPE Vacuum Manifold
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
-
I am surveying SPE extraction device (Visiprep™ SPE Vacuum Manifold), and have 2 questions : [1] if we purchase this device, does it include the pump ? (material #57265) [2] could we use the device for 3cc/60mg, waters SPE?
1 answer-
Helpful?
-
-
Can this unit be used to filter samples through a 0.22µm filter, to separate the solid material and collect the filtrate for further use?
1 answer-
This product is designed to vacuum Solid Phase Extraction (SPE) tubes. The use of a 0.22µm filter for samples has not been confirmed.
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdfHelpful?
-
-
Will the racks suit collection in 50mL centrifuge tubes? My principal concern is in cross contamination between adjacent samples. Does this system prevent or reduce aspiration of fine particles during elution?
1 answer-
The racks for the 24-port model of the Visiprep™ SPE Vacuum Manifold are designed for use with smaller collection vessels. The widest collection vessel recommended for the 24-port model is a small 22.75 mm outer diameter scintillation vial. Since 50 mL centrifuge tubes have a nominal outer diameter of 30 mm, they would not fit this model.
There is no containment system to separate collection vessels during sample processing. The disposable liner from each port can be inserted with slight depth into the respective collection vessel to reduce the likelihood of fine solvent particles being aspirated into the manifold.
Helpful?
-
-
What is the spacing between ports for the manifolds?
1 answer-
Visiprep 24 Port, Product Dimensions
7/8" (0.875") Between valves (center to center)
1 3/8" Between rows (center to center)
All ports on the 12 and 24 vacuum manifolds can only be used simultaneously with 1, 3, and 6mL tubes. In order to use all ports of the manifold with 12, 20, or 60 mL SPE tubes, Long Stem Flow Control Valves are required – see product 57048.Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-
-
HI ON THE Visiprep™ SPE Vacuum Manifold, WHAT is included in initial purchase ( manifold, basin material) etc
1 answer-
The Visiprep™ SPE Vacuum Manifold comes fully assembled, including the basin and collection vessel rack.
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
-
What is the maximum vacuum for the Visiprep Vacuum Manifold, Product 57265?
1 answer-
The maximum vacuum for the Visiprep Vacuum Manifold is 20 inches of Mercury (Hg).
Helpful?
-
-
Can I convert the Visiprep DL 24-port Manifold, Product No. 57265, to a Standard 24-port Manifold, Product No. 57250-U?
1 answer-
Yes, by replacing the manifold lid with Product No. 57251 or by replacing some of the DL ports to standard ports with Product No. 57032 and Product No. 57147-U