Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
2 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 615,00 zł |
Informacje o tej pozycji
615,00 zł
product line
Visiprep™
Quality Segment
packaging
pk of 2
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
compatibility
for use with DL (Disposable Liner) 12- and 24-port models
General description
Application
Visiprep™ manifold flow control valve may also be used for solid phase extraction experiments(SPE) in the determination of biogenic amines in wine samples using high-performance liquid chromatography coupled with fluorescence detector (HPLC).
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description for use with DL (Disposable Liner) 12- and 24-port models, pk of 2 | description for use with standard 12- and 24-port models, pk of 2 | description for use with DL (Disposable Liner) 12- and 24-port models, pk of 24 | description for use with standard 12- and 24-port models, pk of 24 |
| compatibility for use with DL (Disposable Liner) 12- and 24-port models | compatibility for use with standard 12- and 24-port models | compatibility for use with DL (Disposable Liner) 12- and 24-port models | compatibility for use with standard 12- and 24-port models |
| application(s) food and beverages | application(s) food and beverages | application(s) food and beverages | application(s) food and beverages |
| technique(s) solid phase extraction (SPE): suitable | technique(s) solid phase extraction (SPE): suitable | technique(s) solid phase extraction (SPE): suitable | technique(s) solid phase extraction (SPE): suitable |
| packaging pk of 2 | packaging pk of 2 | packaging pk of 24 | packaging pk of 24 |
| product line Visiprep™ | product line Visiprep™ | product line Visiprep™ | product line - |
| Quality Level 100 | Quality Level 100 | Quality Level - | Quality Level - |
Still not finding the right product?
Explore all of our products under Visiprep™ Manifold Flow Control Valve
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?