Przejdź do
57044
Visiprep™ SPE Vacuum Manifold
DL (Disposable Liner), 12-port model
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
1 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
7490,00 zł
Informacje o tej pozycji
UNSPSC Code:
12352200
NACRES:
NB.21
7490,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Złóż zamówienie na produkt na zamówieniePoproś o próbkę
Pomoc techniczna
Pozwól nam pomóc
material
polypropylene tube
Quality Segment
technique(s)
solid phase extraction (SPE): suitable
application(s)
environmental
food and beverages
General description
SPE is a form of chromatography, and, as with other chromatographic techniques, control of flow rate is critical for maintaining reproducible extractions. Unlike other vacuum manifolds, the Visiprep system contains a patented valve system that allows for precise flow control through each SPE tube via rotating, independent, screw-type valves situated in each port within the manifold cover. Visiprep vacuum manifolds allow you to process up to 12 (12-port version) or 24 (24-port version) SPE samples simultaneously. Spacing between ports are slightly smaller on the 24-port versions than the 12-port versions so consideration of tube sizes is recommended.
The Visiprep DL (Disposable Liner) Vacuum Manifold eliminates the possibility of cross-contamination when processing a new sample on the same port. The liner consists of a PP female luer hub that attaches to the SPE, and thin-walled PTFE tubing that is threaded through the SPE port. This ensures that all SPE port and valve surfaces coming in contact with the sample can be replaced following each extraction.
Features and Benefits
  • Patented screw-type valves within each SPE port for precise flow control
  • Glass basin will not dissolve, fog, or discolor when exposed to solvents
  • Legs on cover allow user to easily rest cover on work surface when removed from the manifold
  • Screw-type solvent-resistant vacuum bleed gauge and valve offer better sealing and vacuum control. Valve takes 1/4″ vacuum tubing.
  • PP collection vessel rack accommodates autosampler vials; small scintillation vials (22.75 mm O.D. recommended); 10 and 16 mm test tubes; and 1, 2, 5, and 10 mL volumetric flasks.
Supplied with 100 disposable liners (Part No. 57059).
Legal Information
Visiprep is a trademark of Sigma-Aldrich Co. LLC
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
572755704757254
description
DL (Disposable Liner), 12-port model
description
for use with 3 or 6 mL SPE tubes (includes 4 adapters)
description
for use with standard, 12- and 24-port models (included with 57030-U & 57250-U)), PTFE, pk of 12
description
for use with 24-port model (standard and DL), pk of 2
application(s)
environmental
food and beverages
application(s)
PFAS testing
food and beverages
application(s)
PFAS testing
food and beverages
application(s)
food and beverages
technique(s)
solid phase extraction (SPE): suitable
technique(s)
solid phase extraction (SPE): suitable
technique(s)
solid phase extraction (SPE): suitable
technique(s)
solid phase extraction (SPE): suitable
material
polypropylene tube
material
PTFE frit
material
PTFE
material
-
Quality Level
100
Quality Level
100
Quality Level
100
Quality Level
100
Still not finding the right product?
Explore all of our products under Visiprep™ SPE Vacuum Manifold
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Questions
1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?
    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf
      Helpful?
  2. How is shipping temperature determined? And how is it related to the product storage temperature?
    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
      Helpful?
  3. What is the spacing between ports for the manifolds?
    1 answer
    1. Visiprep 12 Port, Product Dimensions
      1 1/8" (1.14") Between valves (center to center)
      1 5/8" Between rows (center to center)
      All ports on the 12 and 24 vacuum manifolds can only be used simultaneously with 1, 3, and 6mL tubes. In order to use all ports of the manifold with 12, 20, or 60 mL SPE tubes, Long Stem Flow Control Valves are required – see product 57048.