Przejdź do
504335
SPE Tube Adapter
PTFE (with female luer port), for use with 6 mL glass SPE tubes, pk of 24
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
24 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
1610,00 zł
1207,50 zł
Informacje o tej pozycji
UNSPSC Code:
12352200
NACRES:
NB.21
1207,50 zł
Cena katalogowa1610,00 złZaoszczędź 25%
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Złóż zamówienie na produkt na zamówieniePoproś o próbkę
Pomoc techniczna
Pozwól nam pomóc
material
PTFE (with female luer port)
Quality Segment
packaging
pk of 24
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
compatibility
for use with 6 mL glass SPE tubes
General description
Tube adapters serve many purposes. They can be used to stack one SPE tube on top of another to provide different selectivities. A larger empty syringe barrel can be stacked on top of a smaller SPE tube to act as a larger load reservoir. Or, they can serve as an adapter for positive pressure methods (e.g. from a syringe or air/ N2 line).
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
571235712657021
description
PTFE (with female luer port), for use with 6 mL glass SPE tubes, pk of 24
description
configured for AutoTrace Automated Systems, for use with 3mL tubes, pk of 6
description
configured for AutoTrace Automated Systems, for use with 6mL tubes, pk of 6
description
volume 20 mL, pk of 12
application(s)
food and beverages
application(s)
food and beverages
application(s)
food and beverages
application(s)
food and beverages
compatibility
for use with 6 mL glass SPE tubes
compatibility
for use with 3mL tubes, configured for AutoTrace Automated Systems
compatibility
for use with 6mL tubes, configured for AutoTrace Automated Systems
compatibility
-
technique(s)
solid phase extraction (SPE): suitable
technique(s)
solid phase extraction (SPE): suitable
technique(s)
solid phase extraction (SPE): suitable
technique(s)
solid phase extraction (SPE): suitable
material
PTFE (with female luer port)
material
-
material
-
material
polypropylene
packaging
pk of 24
packaging
pk of 6
packaging
pk of 6
packaging
pk of 12
Quality Level
100
Quality Level
100
Quality Level
-
Quality Level
-
Still not finding the right product?
Explore all of our products under SPE Tube Adapter
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)