Merck
23740-U
Supelco
Capillary Cleaving Tool
Fixed Blade Version
About This Item
Kod UNSPSC:
12352200
NACRES:
SB.54
Materiały
sapphire cutting blade
opakowanie
pkg of 1 ea
Opis ogólny
This handy tool makes scalpel-like cuts to polyimide-coated fused silica, leaving no jagged edges to create problems. The industrial sapphire cutting edges remains sharp indefinitely.
Inne uwagi
Fixed blade with a protective cover.
Informacje prawne
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)