Z689521
Jenco™ micrometer for Jenco™ compound microscopes
5 mm scale with 100 divisions
About This Item
Kod UNSPSC:
41191506
Polecane produkty
producent / nazwa handlowa
Jenco
Opis ogólny
You can measure dimensions of the sample within the field of view by inserting one of the Jenco micrometers or reticules into the eyepiece. The reticules are available in multiple configurations to satisfy most needs.
Use the stage micrometer to calibrate the reticule. Its 1 mm scale is divided into 100 divisions.
Use the stage micrometer to calibrate the reticule. Its 1 mm scale is divided into 100 divisions.
Informacje prawne
Jenco is a trademark of Jenco International, Inc.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)