Kluczowe dokumenty
Z375233
Masterflex pump drive
digital standard drive, AC/DC input 190 - 260 V AC
About This Item
Kod UNSPSC:
41100000
producent / nazwa handlowa
Cole Parmer
Opis ogólny
Does not include pump head.
Economy:
unidirectional, lowest priced variable-speed drive, ± 5% speed uniformity. Accepts one EASY-LOAD head
Precision: bidirectional, soft start, 10-turn speed control, ± 1% speed uniformity. Accepts DB9 cable for remote control capabilities
Digital: bidirectional, high-precision digital speed control, ± 0.25% speed uniformity. Displays rpm, flow rate, and % speed. Accepts DB9 cable for remote control capabilities
Informacje prawne
MasterFlex is a trademark of Cole-Parmer Instrument Co.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)