Merck
GE80-6467-79
GE equilibration tubes
About This Item
Kod UNSPSC:
41121800
NACRES:
NB.22
opakowanie
pack of 12 ea
producent / nazwa handlowa
Cytiva 80-6467-79
Cytiva Article No. 80-6467-79
Opis ogólny
Provides a convenient means of equilibrating 18 cm or 24 cm long immobilized pHgradient gel (IPG) strips.