Kluczowe dokumenty
RDHSPACE1
Milli-Q
Spacer for Remote Dispenser
For use with Direct-Q® and Synergy® Water Purification Systems
Wybierz wielkość
1 EA
995,00 zł
995,00 zł
Przewidywany termin wysyłki15 kwietnia 2025Szczegóły
Więcej informacji
Wybierz wielkość
Zmień widok
1 EA
995,00 zł
About This Item
Kod UNSPSC:
41104200
NACRES:
SB.51
995,00 zł
Przewidywany termin wysyłki15 kwietnia 2025Szczegóły
Więcej informacji
opakowanie
pkg of 1 unit
kompatybilność
for use with Direct-Q® UV
for use with Direct-Q®
for use with Synergy® UV
for use with Synergy®
Opis ogólny
The spacer is used when the installation requires the dispenser to be wall-mounted. This accessory increases the distance between the dispenser outlet and the wall. This additional space is required for the dispenser to be positioned right above a sink. It can also be useful when placing large glassware conveniently under the dispenser outlet.
Zastosowanie
Dispensers mounted on a wall can be used with a spacer to gain more depth.
Informacje prawne
DIRECT-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
SYNERGY is a registered trademark of Merck KGaA, Darmstadt, Germany