Z548588
Aldrich® fritted adapter
female joint: ST/NS 29/32, male joint: ST/NS 24/40
About This Item
Kod UNSPSC:
41121815
NACRES:
NB.35
Polecane produkty
Połączenie
female joint: ST/NS29/32
male joint: ST/NS24/40
Opis ogólny
The extra-coarse fritted disc (diameter: 20 mm, porosity A, 145-174 micron) prevents solids from being pulled into the condenser when removing solvents from solid-solvent mixtures. Overall adapter length: 150 mm.
Informacje prawne
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)