Kluczowe dokumenty
Z548553
Sigma-Aldrich
Aldrich® fritted adapter
female joint: ST/NS 24/40, male joint: ST/NS 14/20
About This Item
Kod UNSPSC:
41121815
NACRES:
NB.35
Połączenie
female joint: ST/NS24/40
male joint: ST/NS14/20
Opis ogólny
The extra-coarse fritted disc (diameter: 20 mm, porosity A, 145-174 micron) prevents solids from being pulled into the condenser when removing solvents from solid-solvent mixtures. Overall adapter length: 150 mm.
Informacje prawne
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)