Kluczowe dokumenty
Z173177
Sigma-Aldrich
Aldrich® Schlenk-type adapter
Straight with PTFE stopcock on side-arm, male joint: threaded 14/20, female joint: threaded 14/20
About This Item
Kod UNSPSC:
41121815
NACRES:
NB.43
Połączenie
female joint: threaded14/20
male joint: threaded14/20
Opis ogólny
Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Powiązanie
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Informacje prawne
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)