Merck
SU860098
Supelco
顶空瓶,螺旋盖,圆底(仅小瓶),100个装
volume 20 mL, amber glass vial, thread for 18, O.D. × H 22.5 mm × 75.5 mm, pkg of 100 ea
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41122001
NACRES:
NB.11
材料
amber glass vial
round bottom
特點
closure type screw top vial
包裝
pkg of 100 ea
外徑 × 高度
22.5 mm × 75.5 mm
容積
20 mL
接頭
thread for 18
相容性
for use with CTC PAL (Varian, Gerstel, Atas, Shimadzu)
正在寻找类似产品? 访问 产品对比指南
一般說明
这些样品瓶需要带 18mm 螺纹的专用瓶盖。标准 18-400 螺纹瓶盖不适合该样品瓶。这些样品瓶可用的瓶盖货号为 SU860101(PTFE/硅树脂,1.3mm 厚)、SU860102(PTFE/丁基橡胶,1.6mm 厚)和 SU860103(PTFE/硅树脂,1.5mm 厚)。这些瓶盖使用不锈钢材料并具有磁性。
从最新的版本中选择一种:
分析证书(COA)
Lot/Batch Number
抱歉,我们目前尚未线上提供该产品的COA。
访问文档库
实验方案
通过实验室间试验结果证明了顶空SPME联合GC/MS测定水中VOC方法的优异性能、可靠性和重复性。从检测的灵敏度和选择性来说,采用顶空SPME法的新ISO 17943标准比现有的官方方法更加先进。
High performance, reliability, and reproducibility of HS-SPME in combination with GC/MS for the determination of VOCs in water was proven in an interlaboratory trial. The new ISO 17943 using HS-SPME is an improvement on existing official methods for this determination in terms of sensitivity and selectivity.
High performance, reliability, and reproducibility of HS-SPME in combination with GC/MS for the determination of VOCs in water was proven in an interlaboratory trial. The new ISO 17943 using HS-SPME is an improvement on existing official methods for this determination in terms of sensitivity and selectivity.
High performance, reliability, and reproducibility of HS-SPME in combination with GC/MS for the determination of VOCs in water was proven in an interlaboratory trial. The new ISO 17943 using HS-SPME is an improvement on existing official methods for this determination in terms of sensitivity and selectivity.
查看所有结果