문서
SU860107
Supelco
Vial Rack for SPME
Acrylic Vial Rack for SPME Vial, L × W × H 160 mm × 160 mm × 30 mm, 25 cavities, cavity diam. 24 mm
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41122001
NACRES:
NB.11
material
clear acrylic
특징
25 cavities
포장
pkg of 1 ea
길이 × 너비 × 높이
160 mm × 160 mm × 30 mm
공동 직경
24 mm
Choose from one of the most recent versions:
시험 성적서(COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact 고객 지원 부서