57267
₩204,929
출고 가능일2025년 4월 14일세부사항
1 of 10
Supelco
57020-U
SPE Tube Adapter
57117
Supelclean™ LC-18 SPE Tube
57054
57064
Supelclean™ ENVI™-18 SPE Tube
57012
57030-U
Visiprep™ SPE Vacuum Manifold, PTFE Free
57275
Visiprep™ Large Volume Sampler
54258-U
Large Volume SPE Reservoir
21018-U
Cartridge Adapter for S10, H10, H30 Cartridges
57094
Supelclean™ ENVI-Carb™ SPE Tube
pkg of 6 ea
solid phase extraction (SPE): suitable
environmentalfood and beverages
for use with 12, 20, & 60 mL SPE tubes
Analytical Sample Preparation
Solid Phase Extraction (SPE) and QuEChERS
가장 최신 버전 중 하나를 선택하세요:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 문서 section.
도움이 필요하시면 연락하세요. 고객 지원 부서
문서 라이브러리 방문
1 of 8
54040-U
12mL Si/44%H2SO4/Si SPE Tube
57080-U
Visi-1 Single SPE Tube Processor
CLS9944510
Pyrex® disposable culture tubes, rimless
57179
Empty SPE Tube (no frits)
57273-U
Visiprep™ Large Volume Sampler Tube Adapters, 1/4-28 Threads
52607-U
Discovery® DSC-18 SPE Tube
57161-U
SPE Manifold Gauge/Bleed Valve