주요 문서
EP952010115
Adapter for Eppendorf® UVette®, for photometer/spectrophotometer with light path height
size 15 mm
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121815
제품 EP952010115은(는)귀하의 국가에서 현재 판매되지 않습니다. 고객지원팀으로 연락바랍니다.
제조업체/상표
Eppendorf® 952010115
크기
15 mm
유사한 제품을 찾으십니까? 방문 제품 비교 안내
관련 카테고리
일반 설명
This Eppendorf adaptor is for use with photometers/spectrophotometers which provides 10mm light centered height specifically for the Eppendorf UVette Disposable Cuvette (sold separately). The UVette is ideal for use with the Eppendorf BioPhotometer. In other photometers and/or spectrophotometers, the UVette is inserted with the aid of an adapter. This adapter aligns the UVette to the corresponding height of the optical light path in the instrument. Adapters are also available for use with other common spectrophotometers. Additional adapters for light center heights of 8mm or 15mm are also available (952010107 for 8mm and 952010115 for 15mm). An adapter list is available that provides you with an overview of the correct adapter for use with Eppendorf′s UVette in devices from other manufacturers. No adapter is required for use in Eppendorf′s photometers. The adapters′ compatibility with the devices from other manufacturers has been jointly tested by Eppendorf, the device manufacturers and our customers. Size: 10mm.
법적 정보
Eppendorf is a registered trademark of Eppendorf AG
UVette is a registered trademark of Eppendorf AG
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)