GCによるtrans-1,2-シクロヘキサンジオールエナンチオマーの分析、Astec® CHIRALDEX™ B-PH
CONDITIONS
column
Astec CHIRALDEX B-PH, 30 m x 0.25 mm I.D., 0.12 μm (71023AST)
oven
120 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
sample
peaks 1 & 2: trans-1,2-cyclohexanediol enantiomers