主要文書
1.17333
Millipore
MAS-100 Atmos® Hardware Key Set (5x)
for use with MAS-100 Atmos®
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41103403
NACRES:
NA.74
現在、価格および在庫状況を閲覧できません。
詳細
Compressed gas sampling
特徴
electronic
メーカー/製品名
MAS-100 Atmos®
アプリケーション
compressed gas monitoring
cosmetics
food and beverages
pharmaceutical
適合性
for use with MAS-100 Atmos®
詳細
The MAS-100 Atmos® Hardware Key Set (5x) is designed uniquely for each individual user role. It is available in different colors for easy identification of each user.
The hardware key is a data carrier based on single-wire technology. It contains the login data for accessing the instrument function according to the user management access levels.
アプリケーション
The MAS-100 Atmos® Hardware Key Set (5x) is used for access restricted login to operate the instrument.
特徴および利点
  • 21 CFR part 11 compliant individual login on MAS-100 Atmos®
  • Re-writable
法的情報
MAS-100 Atmos is a registered trademark of MBV AG